Adapters, Flow Control, Chromatography, Rodaviss® Joints, Chemglass
Supplier: Chemglass
CG-1195-10 CG-1195-11
80066-896EA 405.27 CAD
80066-896 80066-898
Adapters, Flow Control, Chromatography, Rodaviss® Joints, Chemglass
Adapters Adapters, Joints with Tube Connectors
Flow adapter for use with CG-1192 and CG-1193 chromatography columns along with CG-1194 reservoirs.
  • Flow control adapter, chromatography, Rodaviss® joint
  • Adapter has a 0 to 4 mm chem-cap metering plug