Buchner Filter Funnels with Inner Joints, Improved, Chemglass
Supplier: Chemglass
CG-1406-E-04 CG-1406-E-14 CG-1406-E-15 CG-1406-E-16 CG-1406-E-01 CG-1406-E-02 CG-1406-E-03 CG-1406-E-05 CG-1406-E-06 CG-1406-E-07 CG-1406-E-08 CG-1406-E-09 CG-1406-E-11 CG-1406-E-12 CG-1406-E-13
80068-514EA 446.32 CAD
80068-514 80068-534 80068-536 80068-538 80068-508 80068-510 80068-512 80068-516 80068-518 80068-520 80068-522 80068-524 80068-526 80068-528 80068-530 80068-532
Buchner Filter Funnels with Inner Joints, Improved, Chemglass
Funnels Filtration Funnels Büchner Funnels
Buchner style filter funnel features a lower standard taper inner joint.
  • Serrated tubulation for connection to a vacuum source
  • Disc diameters and height above the disc are the same as Buchner funnel
  • Improved design incorporates a delivery stem being sealed within the inner joint to prevent breakage
  • Funnel is supplied with a 3" piece of PTFE tubing that slides over the delivery stem
The tubing permits the funnel to be used during filtrations at higher vacuums without having the fluid pulled into the vacuum source. Overall height is without the PTFE tubing.