Funnels, Addition, PTFE Stopcocks, Chemglass
Supplier: Chemglass
CG-1702-06 CG-1702-07 CG-1702-08 CG-1702-09 CG-1702-11 CG-1702-01 CG-1702-05 CG-1702-10 CG-1702-12 CG-1702-13 CG-1702-14 CG-1702-15 CG-1702-16 CG-1702-17
80070-514EA 929.43 CAD
80070-514 80070-516 80070-518 80070-520 80070-524 80070-504 80070-512 80070-522 80070-526 80070-528 80070-530 80070-532 80070-534 80070-536
Funnels, Addition, PTFE Stopcocks, Chemglass
Funnels Dropping Funnels
Funnels feature a standard taper joint.
  • Standard taper top outer joint and lower inner joint, both being the same size
  • With pressure equalizing arm and PTFE stopcock
Funnels with 14/20 and 19/22 joints are supplied complete with a stopper.
Ordering information: For replacement stoppers see CG-3000.