Peptide Synthesis Vessels, Solid Phase, T-Bore PTFE Stopcocks, Chemglass
Supplier: Chemglass
CG-1864-01 CG-1864-02 CG-1864-03 CG-1864-04 CG-1864-05
80071-368EA 593.86 CAD
80071-368 80071-370 80071-372 80071-374 80071-376
Peptide Synthesis Vessels, Solid Phase, T-Bore PTFE Stopcocks, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Solid phase peptide synthesis vessel having a medium porosity, fritted glass resin support.
  • Top of the vessel has a GL thread
  • Supplied complete with a PTFE lined PBT screw cap
Lower stopcock is a 'T' bore 2 mm PTFE to allow nitrogen to be used to upwardly agitate the resin beads while shaking. The O.D. of both lower stems is 8 mm.