Condensers, Reflux, High Efficiency, Removable Hose Connections, Chemglass
Supplier: Chemglass
CG-1217-HC-25 CG-1217-HC-20 CG-1217-HC-21
16002-688EA 1662.44 CAD
16002-688 16002-682 16002-684
Condensers, Reflux, High Efficiency, Removable Hose Connections, Chemglass
Condensers
Compact high efficiency reflux condenser similar to CG-1217 but with detachable hose connections.
  • Connections are made of polypropylene
  • Provide a safe and easy way of connecting and disconnecting condensers
  • Flexible tubing to the condenser
Condensing area is the same as CG-1217.
Delivery information: Supplied complete with two GL open top screw caps and two straight polypropylene hose connections.