White SMS Coveralls with Attached Hood, Elastic Wrist and Ankle, International Enviroguard™
Supplier: International Enviroguard
2225-M 2225-L 2225-XL 2225-2XL 2225-3XL 2225-4XL
83007-072CS 293.2 CAD
83007-072 83007-074 83007-076 83007-078 83007-080 83007-082
White SMS Coveralls with Attached Hood, Elastic Wrist and Ankle, International Enviroguard™
Disposable Garments
SMS Coverall provide an excellent particulate barrier along with protection from light liquid splashes and sprays.
  • SMS fabric is strong yet breathable to keep users cool and help avoid heat stress
  • Storm flap over zipper adds an extra layer of protection
  • Attached hood with elastic wrists and ankles for snug, secure fit
Three-layer SMS is lightweight and breathable. Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric. Elastic back stretches with user for more comfort and better tear resistance.
RecommendedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Closure type Zip closure
Gender
Garment application Food,Industrial,General Purpose,Laboratory,Pharmaceutical