White SMS Coveralls, Elastic Wrist and Ankle, International Enviroguard™
Supplier: International Enviroguard
2223-L 2223-XL 2223-2XL 2223-3XL 2223-4XL
83007-062CS 270.5 CAD
83007-062 83007-064 83007-066 83007-068 83007-070
White SMS Coveralls, Elastic Wrist and Ankle, International Enviroguard™
Disposable Garments
SMS Coveralls provide an excellent particulate barrier along with protection from light liquid splashes and sprays.
  • SMS fabric is strong yet breathable to keep users cool and help avoid heat stress
  • Storm flap over zipper adds an extra layer of protection
  • Elastic wrists and ankles for snug, secure fit
Three-layer SMS fabric is lightweight and breathable. Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric. Elastic back stretches with user for more comfort and better tear resistance.
Recommended applications include asbestos and lead abatement, mold remediation, manufacturing, assembly operations, and more.