Condensers, Reflux, Coil Type, Removable Hose Connections, Chemglass
Supplier: Chemglass
CG-1213-J-HC01 CG-1213-J-HC02
30622-516EA 1271.42 CAD
30622-516 30622-518
Condensers, Reflux, Coil Type, Removable Hose Connections, Chemglass
Condensers
Coil style reflux condenser feature a larger diameter jacket.
  • Hose connections are on the bottom of the condenser
  • Makes it easier to connect hose/tubing connections are GL14 polypropylene hose barbs and screw caps
Coil style reflux condenser with a larger diameter jacket and coil, thereby providing maximum cooling capacity, while being shorter in length. GL hose connections provide a safe and easy way of connecting flexible tubing to a condenser. Jacket has a O.D. of 41 mm while the coil O.D. is 8 mm. Excellent for use with high vapor pressureMore Product Information