Addition Funnels, Graduated with PTFE Stopcock, Chemglass
Supplier: Chemglass
CG-1712-05 CG-1712-07 CG-1712-08 CG-1712-09 CG-1712-12 CG-1712-14 CG-1712-15 CG-1712-20 CG-1712-21 CG-1712-22
80070-644EA 1014.36 CAD
80070-644 80070-648 80070-650 80070-652 80070-658 80070-662 80070-664 80070-674 80070-676 80070-678
Addition Funnels, Graduated with PTFE Stopcock, Chemglass
Funnels Dropping Funnels
Cylindrical style addition funnels feature a PTFE stopcock.
  • Funnels with 14/20 and 19/22 joints are complete with a stopper
Ordering information: Replacement stoppers (CG-3000) can be ordered separately.