Flasks, Heavy Wall, Round Bottom with Three Necks, Jacketed, Chemglass
Supplier: Chemglass
CG-1538-01 CG-1538-02 CG-1538-04 CG-1538-05 CG-1538-07 CG-1538-08 CG-1538-10 CG-1538-14 CG-1538-15 CG-1538-16 CG-1538-17
80061-322EA 2117.53 CAD
80061-322 80061-324 80061-328 80061-330 80061-334 80061-336 80061-340 80061-348 80061-350 80061-352 80061-354
Flasks, Heavy Wall, Round Bottom with Three Necks, Jacketed, Chemglass
Flasks Round Bottom Flasks
Round bottom, heavy wall, three-neck flasks feature a standard taper.
  • Entire body of the flask is jacketed
  • Has two hose connections for circulation of heating or cooling liquid
Standard taper outer joints on the center and side necks.