Flasks, Heavy Wall, Round Bottom, 4-Necks, Chemglass
Supplier: Chemglass
CG-1531-03 CG-1532-05 CG-1532-06 CG-1532-07 CG-1532-08 CG-1532-09 CG-1532-11 CG-1532-13 CG-1532-15 CG-1532-32 CG-1532-33 CG-1532-35 CG-1532-38 CG-1532-47 CG-1532-53 CG-1532-54 CG-1532-57 CG-1531-01 CG-1531-05 CG-1531-06 CG-1531-07 CG-1531-09 CG-1532-01 CG-1532-03 CG-1532-45 CG-1532-46
80069-810EA 348.6 CAD
80069-810 80069-830 80069-832 80069-834 80069-836 80069-838 80069-842 80069-846 80069-850 80069-856 80069-858 80069-862 80069-868 80069-886 80069-896 80069-898 80069-900 80069-806 80069-814 80069-816 80069-818 80069-822 80069-826 80069-828 80069-882 80069-884
Flasks, Heavy Wall, Round Bottom, 4-Necks, Chemglass
Flasks Round Bottom Flasks
Flasks feature a round bottom.
  • Heavy wall four neck flasks
  • Standard taper outer joints on the center
  • 20° angled side necks