Adapters, Rotary Evaporator, Microwave Vials, PTFE, Chemglass
Supplier: Chemglass
CG-1318-A-10 CG-1318-A-12
89041-338EA 557.45 CAD
89041-338 89041-340
Adapters, Rotary Evaporator, Microwave Vials, PTFE, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters feature a standard taper joint.
  • PTFE adapter allows convenient connection
  • Connecting microwave vial to a rotary evaporator bump
  • Trap having a standard taper joint size
Lower portion incorporates a threaded black Acetal compression cap that is slotted to accept the flange fitting on the vial.