Condensers, Reflux, 300 mm, Removable Hose Connections, Chemglass
Supplier: Chemglass
CG-1213-L-HC01 CG-1213L-HC-02 CG-1213L-HC-03
16002-666EA 815.99 CAD
16002-666 16002-668 16002-670
Condensers, Reflux, 300 mm, Removable Hose Connections, Chemglass
Condensers
Condensers feature two GL open top screw caps and are a great a value.
  • Connections are made of polypropylene
  • Provide a safe and easy way of connecting and disconnecting to the condenser
  • Flexible tubing to the condenser
Coil style reflux condenser with a large diameter jacket and coil, with detachable hose connections.
Delivery information: Supplied complete with two GL open top screw caps and two bent polypropylene hose connections.