Condensers, Friedrichs, Serrated Hose Connection, Chemglass
Supplier: Chemglass
CG-1211-01 CG-1211-03
80067-270EA 688.23 CAD
80067-270 80067-272
Condensers, Friedrichs, Serrated Hose Connection, Chemglass
Condensers
Condensers with a serrated hose connection for use with a drying tube.
  • Condenser, Friedrichs with overall height: 330 mm
  • This style condenser is used primarily in Soxhlet extractions or in Reflux mode
Hose connections have an O.D. of 10 mm at the middle serration.