Reaction Vessel Lids, 5-Necks, 10° Angled Side Necks, Chemglass
Supplier: Chemglass
CG-1946-D-01 CG-1946-D-03 CG-1946-D-05 CG-1946-D-11
30622-552EA 1221.15 CAD
30622-552 30622-554 30622-556 30622-558
Reaction Vessel Lids, 5-Necks, 10° Angled Side Necks, Chemglass
Lids Reaction Vessel Lids
Reaction vessel lids feature five necks.
  • Five neck vessel lid to fit the reaction vessels
  • Two side necks are vertical and two necks have a 10° degree angle
Angled side neck is designed for use with angled distillation adapters. Lid has a flat ground surface allowing it to be greased or used with an O-ring.