Flasks, Heavy Wall, Round Bottom, 4-Necks, Chemglass
Supplier: Chemglass
CG-1533-06 CG-1533-07 CG-1533-08 CG-1533-10 CG-1533-11 CG-1533-12 CG-1533-15 CG-1533-16 CG-1533-19 CG-1533-20 CG-1533-21 CG-1533-01 CG-1533-02 CG-1533-03 CG-1533-04 CG-1533-05
80069-912EA 665.46 CAD
80069-912 80069-914 80069-916 80069-918 80069-920 80069-922 80069-926 80069-928 80069-934 80069-936 80069-938 80069-902 80069-904 80069-906 80069-908 80069-910
Flasks, Heavy Wall, Round Bottom, 4-Necks, Chemglass
Flasks Round Bottom Flasks
Flasks feature a round bottom.
The third neck is a 10/30 outer joint for use with a CG-3506, 10/30 jointed thermometer. Flasks with 29/26 center necks have a groove tooled just below the grind to aid in the clamping of the flask.