Flasks, Evaporating, Large Scale, Flanged, Compatible with Buchi, Chemglass
Supplier: Chemglass
CG-1337-08 CG-1337-09 CG-1337-02 CG-1337-05 CG-1337-06 CG-1337-07 CG-1337-14
47733-792EA 1700.03 CAD
47733-792 47733-794 80060-980 80060-986 80060-988 80060-990 89028-438
Flasks, Evaporating, Large Scale, Flanged, Compatible with Buchi, Chemglass
Flasks Rotary Evaporator Flasks
Flasks are compatible with Buchi large scale rotary evaporators.
  • Available plain or plastic coated to prevent injury from broken glass while preventing spillage of hazardous materials
  • Flasks to fit large scale rotary evaporators
  • Flasks have a heavy wall for vacuum applications and are balanced to prevent run-out during rotation
To select the proper flask for your evaporator:
Determine the size flange required from the chart below
Match the overall height with the capacity required