Vapor Tubes, Compatible with Buchi Rotary Evaporators, Chemglass
Supplier: Chemglass
CG-1317-G-10 CG-1317-G-12
89209-328EA 1029.25 CAD
89209-328 89209-330
Vapor Tubes, Compatible with Buchi Rotary Evaporators, Chemglass
Tubes Vapour Tubes
Replacement vapor tubes are compatible with Buchi rotary evaporators.
  • Thread is made for use with standard Combi-Clip
Buchi Part # 23737 is equal to 89209-328
Buchi Part # 23738 is equal to 89209-330
* Length 'A', refer to image.
Ordering information: Not supplied with Combi-clip.