Condensers, Allihn, Removable Hose Connections, Chemglass
Supplier: Chemglass
CG-1206-HC-01 CG-1206-HC-05
47733-776EA 502.4 CAD
47733-776 47733-778
Condensers, Allihn, Removable Hose Connections, Chemglass
Condensers
Allihn style reflux condenser similar to CG-1206 but with detachable hose connections.
  • Connections are made of polypropylene
  • Provide a safe and easy way of connecting and disconnecting to the condenser
  • Flexible tubing to the condenser
Hose connections have an I.D. of 4 mm.
Ordering information: Supplied complete with two GL 14 open top screw caps. See CG-198 and CG-195 for replacement GL fittings.