Buchner Funnels, Perforated Plates, Chemglass
Supplier: Chemglass
CG-1402-P-02 CG-1402-P-03 CG-1402-P-01
10170-344EA 259.74 CAD
10170-344 10170-346 10170-348
Buchner Funnels, Perforated Plates, Chemglass
Funnels Filtration Funnels Büchner Funnels
Buchner funnels having a perforated plate rather than a fritted disc.
Funnels accept filter paper size as listed.