Flasks, Heavy Wall, Round Bottom, 3-Necks, Jacketed, Morton Indents, Chemglass
Supplier: Chemglass
CG-1538-M-20 CG-1538-M-23 CG-1538-M-24 CG-1538-M-26 CG-1538-M-27 CG-1538-M-31 CG-1538-M-32 CG-1538-M-35 CG-1538-M-36
80061-356EA 2206.97 CAD
80061-356 80061-362 80061-364 80061-368 80061-370 80061-378 80061-380 80061-386 80061-388
Flasks, Heavy Wall, Round Bottom, 3-Necks, Jacketed, Morton Indents, Chemglass
Flasks Round Bottom Flasks
Round bottom, heavy wall, three-flask with standard taper outer joints on the center and side necks.
  • Entire body of the flask is jacketed
  • Has two hose connections for circulation of heating or cooling liquid
The indentations increase agitation of the contents of the flask during stirring.