Condensers, C Assembly, For Rotary Evaporators, Plastic Coated, Chemglass
Supplier: Chemglass
CG-1333-11 CG-1333-10
80060-954EA 2055.34 CAD
80060-954 80060-952
Condensers, C Assembly, For Rotary Evaporators, Plastic Coated, Chemglass
Condensers
Two piece cold trap style Condenser for use with rotary evaporators.
  • Condenser has a 50 mm flange on the side
  • A 35/20 lower ball joint
  • A 19/38 stopcock plug
  • Removable hose connections using GL-14 threads
Condenser is supplied complete with PTFE centering ring along with plastic caps and serrated hose connections. Plastic coating is on outer portion of the condenser only.
Order Now
SPECIFICATIONS
Environmentally Preferable