Adapters, Rotary Evaporators, Vials, Chemglass
Supplier: Chemglass
CG-1318-41 CG-1318-42
89028-422EA 470.47 CAD
89028-422 89028-424
Adapters, Rotary Evaporators, Vials, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters permit the connection of standard laboratory dram sample vials directly to the vapor tube or a bump trap of a rotary evaporator.
  • Adapters feature a top outer joint
Adapters have a top outer joint of the listed size and a 20 to 400 G.P.I. thread at the bottom. A PTFE-lined, polypropylene, double-threaded connector attaches the vial to the adapter.