Condensers, Reflux, Removable Hose Connections, Chemglass
Supplier: Chemglass
CG-1213-HC-09 CG-1213-HC-01 CG-1213-HC-03
47733-788EA 816.42 CAD
47733-788 47733-784 47733-786
Condensers, Reflux, Removable Hose Connections, Chemglass
Condensers
Reflux Condenser similar to CG-1213 but with detachable hose connections.
  • Connections are made of polypropylene and provide a safe and easy way of connecting disconnecting flexible tubing to the condenser
Hose connections have an O.D. of 8 mm and an I.D. of 4 mm.
Delivery information: Supplied complete with two GL-14 open top screw caps. See CG-198 for replacement GL fittings.