Condensers, Concentrators, Distillation Heads, Distilling, Chemglass
Supplier: Chemglass
CG-1208-20 CG-1208-21
80067-228EA 1308.44 CAD
80067-228 80067-230
Condensers, Concentrators, Distillation Heads, Distilling, Chemglass
Distillation Components Distillation Heads
Vapors rise to the top of inner tube then travel downward and condense in this highly efficient concentrator.
  • Condensate drains through the side tube which has a standard taper inner joint the same size as the lower inner uptake joint
Serrated hose connection for connecting to vacuum source. Hose connections for both the cooling jacket and vacuum source have an O.D. of 10 mm.
Ordering information: Available with detachable hose connections on a special order basis.