Liquid-Liquid Extractor, Continuous, Round Bottom, Chemglass
Supplier: Chemglass
CG-1366-01
80068-150EA 1776.54 CAD
80068-150
Liquid-Liquid Extractor, Continuous, Round Bottom, Chemglass
Extractors Liquid-Liquid Extractors
Round bottom design has the return tube sealed into the bottom of the extractor for more efficient extractions.
  • For samples of 1 liter plus approximately 200 ml of extracting solvent
  • Extractor has a 45/50 top outer joint and a 24/40 inner joint on the uptake tube
Ordering information: Flask and condenser must be ordered separately.