Flasks, Tapered, European Style, 3-Necks, Chemglass
Supplier: Chemglass
CG-1570-10 CG-1570-12 CG-1570-01 CG-1570-02 CG-1570-03 CG-1570-04 CG-1570-05 CG-1570-06 CG-1570-08
80070-318EA 876.06 CAD
80070-318 80070-322 80070-300 80070-302 80070-304 80070-306 80070-308 80070-310 80070-314
Flasks, Tapered, European Style, 3-Necks, Chemglass
Flasks Round Bottom Flasks
Flasks feature a european design with three necks.
  • European style flask with tapered walls and a shallow hemispherical bottom
  • For more efficient mechanical or magnetic stirring
  • Side neck outer angled 20°
Standard taper outer joints on center and side necks.