Human Recombinant TLR-4 (from HEK293 Cells)
Supplier: Peprotech
160-06-10UG 160-06-50UG 160-06-100UG 160-06-250UG 160-06-500UG 160-06-1MG
76303-748EA 196.27 CAD
76303-748 76303-750 76303-752 76303-754 76303-756 76303-758
Human Recombinant TLR-4 (from HEK293 Cells)
Proteins and Peptides
Recombinant Human TLR-4 is a 69.3 kDa glycoprotein containing 609 amino acid residues of the TLR-4 extracellular domain. As a result of glycosylation, Recombinant Human TLR-4 migrates with an apparent molecular mass of approximately 90-100 kDa by SDS-PAGE gel, under reducing and non-reducing conditions.