Human Recombinant PlGF-2 (from E. coli)
Supplier: Peprotech
100-56A-5UG 100-56A-25UG 100-56A-100UG 100-56A-250UG 100-56A-500UG 100-56A-1MG
76303-698EA 196.27 CAD
76303-698 76303-700 76303-702 76304-080 76304-082 76304-084
Human Recombinant PlGF-2 (from E. coli)
Proteins and Peptides
PlGF-2 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-2 is expressed in umbilical vein endothelial cells and placenta. It signals through the VEGFR-1/FLT1 receptor, and stimulates endothelial cell proliferation and migration. PlGF-2 also signals through Neuropilin (NP-1), and can bind with high affinity to heparin. Recombinant Human PlGF-2 is a 34.6 kDa disulfide-linked homodimeric protein of two 152 amino acid polypeptide chains.