Human Recombinant IL-35 (from HEK293 Cells)
Supplier: Peprotech
200-37-50UG 200-37-100UG 200-37-250UG 200-37-500UG
76303-800EA 1570.61 CAD
76303-800 76303-802 76303-804 76303-806 76303-808
Human Recombinant IL-35 (from HEK293 Cells)
Proteins and Peptides
Interleukin-35 is a glycosylated, heterodimeric protein consisting of the p35 subunit from IL-12 (IL-12alpha) and the beta subunit from IL-27 (EBI3). IL-35 can be expressed by regulatory T-cells (Tregs), macrophages, and certain trophoblast and dendritic cells. It is induced in response to inflammation, and generally acts as an inflammation suppressor. IL-35 suppresses inflammation by exerting multiple activities, including the induction of regulatory T-cells and the suppression of Th17 cells. Recombinant Human IL-35 produced from HEK293 cells is a glycosylated protein, primarily heterodimeric, that migrates as a diffuse band centered at an apparent molecular weight of about 60 kDa by SDS-PAGE analysis under non-reducing conditions. Recombinant Human IL-35 expressed in HEK293 cells contains 406 amino acid residues and has a calculated molecular weight of 45.8 kDa.