Human Recombinant ICOS (from CHO cells)
Supplier: Peprotech
310-39-10UG 310-39-50UG 310-39-100UG 310-39-250UG 310-39-500UG 310-39-1MG
76303-910EA 196.27 CAD
76303-910 76303-912 76303-914 76303-916 76303-918 76303-920
Human Recombinant ICOS (from CHO cells)
Proteins and Peptides
PeproTech's CHO cell-derived Recombinant Human ICOS Fc is a glycosylated, homodimer 79.4 kDa of 706 amino acid residues whose monomer consists of the 120-amino-acid-length extracellular portion of ICOS fused to the 231-amino-acid-length Fc portion of human IgG1 by two glycines. The calculated molecular weight of Recombinant Human ICOS Fc dimer is 79.4 kDa; however, due to glycosylation, it migrates at an apparent molecular weight of approximately 40-45 kDa by SDS-PAGE analysis under reducing conditions.