Rat Recombinant GDNF (from E. coli)
Supplier: Peprotech
AF-450-51-2UG AF-450-51-50UG AF-450-51-500UG
75999-672EA 266.99 CAD
75999-672 75999-676 75999-682
Rat Recombinant GDNF (from E. coli)
Proteins and Peptides
GDNF is a disulfide-linked, homodimeric neurotrophic factor structurally related to Artemin, Neurturin and Persephin. These proteins belong to the cysteine-knot superfamily of growth factors that assume stable dimeric protein structures. GDNF signals through a multicomponent receptor system, composed of a RET and one of the four GFRalpha (alpha1-alpha4) receptors. GDNF specifically promotes dopamine uptake and survival, and morphological differentiation of midbrain neurons. Using a Parkinson's disease mouse model, GDNF has been shown to improve conditions such as bradykinesia, rigidity, and postural instability. The functional rat GDNF ligand is a disulfide-linked homodimer consisting of two 15 kDa polypeptide chains called monomers. Each monomer contains seven conserved cysteine residues, including Cys-101, which is used for inter-chain disulfide bridging, and others that are involved in the intramolecular ring formation known as the cysteine-knot configuration. The calculated molecular weight of Recombinant Rat GDNF is 30.0 kDa.
  • Animal-free