Human Recombinant GRO-alpha/MGSA (CXCL1) (from E. coli)
Supplier: Peprotech
AF-300-11-5UG AF-300-11-25UG AF-300-11-100UG AF-300-11-250UG AF-300-11-500UG AF-300-11-1MG
76304-148EA 249.72 CAD
76304-148 76304-150 76304-152 76304-154 76304-156 76304-158
Human Recombinant GRO-alpha/MGSA (CXCL1) (from E. coli)
Proteins and Peptides
All three isoforms of GRO are CXC chemokines that can signal through the CXCR1 or CXCR2 receptors. The GRO proteins chemoattract and activate neutrophils and basophils. GRO/MGSA also stimulates mitogenesis in certain human melanoma cells. Recombinant Human GRO-alpha/MGSA is a 7.8 kDa protein consisting of 73 amino acids, including the 'ELR' motif common to the CXC chemokine family that binds to CXCR1 or CXCR2.
  • Animal-free