Human Recombinant FGF-19 (from E. coli)
Supplier: Peprotech
AF-100-32-5UG AF-100-32-25UG AF-100-32-1MG
76081-526EA 266.99 CAD
76081-526 76081-528 76081-536
Human Recombinant FGF-19 (from E. coli)
Proteins and Peptides
The FGF family plays central roles during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-19, a member of the FGF family, is a high-affinity heparin dependent ligand for FGFR4. FGF-19 is expressed during brain development and embryogenesis. Recombinant Human FGF-19 is a 21.8 kDa protein containing 195 amino acid residues.
  • Animal-free