Human Recombinant BMP-13 (from E. coli)
Supplier: Peprotech
AF-120-04-100UG AF-120-04-250UG AF-120-04-500UG
76077-278EA 922.8 CAD
76077-278 76077-280 76077-470
Human Recombinant BMP-13 (from E. coli)
Proteins and Peptides
BMP-13 is expressed in hypertrophic chondrocytes during embryonic development of long bones. Continued postnatal expression of BMP-13 in articular cartilage suggests that it plays a regulatory role in the growth and maintenance of articular cartilage. Adenovirus-mediated BMP-13 gene transfer to rabbit bone marrow stem cells have been reported to augment periosteal repair of osteochondral defects. The functional form of BMP-13/CDMP-2 is a disulfide-linked homodimer of two 120 amino-acid polypeptide chains. This 27.5 kDa protein is obtained by proteolytic processing of a biologically inactive precursor protein of 97.7kDa. Recombinant Human BMP-13/CDMP-2 is a 27.0 kDa homodimeric disulfide-linked protein consisting of two 120 amino acid polypeptide chains.
  • Animal-free