Anti-IgG Rabbit Polyclonal Antibody (Biotin) Amphiregulin

Supplier: Peprotech
500-P322BT-50UG 500-P322BT-25UG
76304-224EA 642.3 CAD
76304-224 76304-226 76304-314
Anti-IgG Rabbit Polyclonal Antibody (Biotin) Amphiregulin
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR