Anti-IgG Rabbit Polyclonal Antibody (Biotin) MMP-3

Supplier: Peprotech
500-P324BT-25UG 500-P324BT-50UG
76304-180EA 502.16 CAD
76304-180 76304-182 76304-184
Anti-IgG Rabbit Polyclonal Antibody (Biotin) MMP-3
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR