Anti-IgG Rabbit Polyclonal Antibody (Biotin) VAP-1

Supplier: Peprotech
500-P326BT-25UG 500-P326BT-50UG
76304-204EA 502 CAD
76304-204 76304-206 76304-208
Anti-IgG Rabbit Polyclonal Antibody (Biotin) VAP-1
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR