Anti-IgG Rabbit Polyclonal Antibody (Biotin) Resistin

Supplier: Peprotech
500-P183BT-50UG 500-P183BT-25UG
76303-984EA 642.3 CAD
76303-984 76303-986 76304-110
Anti-IgG Rabbit Polyclonal Antibody (Biotin) Resistin
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR