Anti-IgG Rabbit Polyclonal Antibody (Biotin) sCD14

Supplier: Peprotech
500-P320BT-25UG 500-P320BT-50UG
76304-054EA 502.16 CAD
76304-054 76304-056 76304-058
Anti-IgG Rabbit Polyclonal Antibody (Biotin) sCD14
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR