Anti-IgG Rabbit Polyclonal Antibody sCD14

Supplier: Peprotech
500-P320-50UG 500-P320-100UG 500-P320-1MG
76304-048EA 502.16 CAD
76304-048 76304-050 76304-052
Anti-IgG Rabbit Polyclonal Antibody sCD14
Antibodies
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR