Anti-IgG Rabbit Polyclonal Antibody (Biotin) Anti-Human Osteopontin

Supplier: Peprotech
500-P314BT-25UG 500-P314BT-50UG
76304-024EA 502.16 CAD
76304-024 76304-026
Anti-IgG Rabbit Polyclonal Antibody (Biotin) Anti-Human Osteopontin
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR