Anti-IgG Rabbit Polyclonal Antibody (Biotin) Anti-Murine LIGHT

Supplier: Peprotech
500-P308BT-25UG 500-P308BT-50UG
76304-018EA 502.16 CAD
76304-018 76304-020 76304-022
Anti-IgG Rabbit Polyclonal Antibody (Biotin) Anti-Murine LIGHT
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR