Anti-IgG Rabbit Polyclonal Antibody LIGHT

Supplier: Peprotech
500-P308-50UG 500-P308-100UG 500-P308-1MG
76304-012EA 502.16 CAD
76304-012 76304-014 76304-016
Anti-IgG Rabbit Polyclonal Antibody LIGHT
Antibodies
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR