Anti-IgG Rabbit Polyclonal Antibody Adiponectin

Supplier: Peprotech
500-P239-50UG 500-P239-100UG 500-P239-1MG
76303-988EA 502.16 CAD
76303-988 76303-990 76303-992
Anti-IgG Rabbit Polyclonal Antibody Adiponectin
Antibodies
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR