Anti-IgG Rabbit Polyclonal Antibody sIL-2 Receptor alpha

Supplier: Peprotech
500-P22R-50UG 500-P22R-100UG 500-P22R-1MG
76303-892EA 502 CAD
76303-892 76303-894 76303-896
Anti-IgG Rabbit Polyclonal Antibody sIL-2 Receptor alpha
Antibodies
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR