Anti-IgG Rabbit Polyclonal Antibody (Biotin) RELMalpha

Supplier: Peprotech
500-P214BT-25UG 500-P214BT-50UG
10776-948EA 536.88 CAD
10776-948 10776-950
Anti-IgG Rabbit Polyclonal Antibody (Biotin) RELMalpha
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR