Anti-IgG Rabbit Polyclonal Antibody Oncostatin M

Supplier: Peprotech
500-P30-50UG 500-P30-100UG
10775-158EA 536.88 CAD
10775-158 10775-160
Anti-IgG Rabbit Polyclonal Antibody Oncostatin M
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Oncostatin M. Anti-Human Oncostatin M-specific antibody was purified by affinity chromatography employing an immobilized Human Oncostatin M matrix.

Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR