CleanTex™ Hydrosorb I Polyester/Cellulose Non-Woven Wipes, Advantus
Supplier: Advantus
CT604 CT609
33344-038PK 211.64 CAD
33344-038 33344-042
CleanTex™ Hydrosorb I Polyester/Cellulose Non-Woven Wipes, Advantus
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
Combines the highly absorbent properties of a natural fiber with the cleanliness and strength of a synthetic wipe to clean up aqueous spills, precision components, and lab equipment.
  • Hydrosorb I wipes are fabricated from a hydroentangled, non-woven blend of 55% cellulose and 45% polyester
  • CleanTex™ Hydrosorb I wipes offer excellent strength, both wet and dry
  • Compatible with most solvents
  • No chemical binders or additives
  • Low particle counts and extractable levels make these wipes virtually lint-free
  • Autoclavable
These CleanTex™ Hydrosorb I Wipes for superior durability and absorbency. Hydrosorb I is ideal for general wiping and spill control in controlled environments. Designed for general-purpose wiping where economy and cleanliness are of critical importance. These wipes are ideal for applications where high absorbency and low extractable levels are of paramount concern.