Electrophoresis Buffer Stock Powders
Supplier: Ward's
EB1000_WARDS EB1100_WARDS
470318-976BX 359.3 CAD
470318-976 470318-980 470326-130 470326-132 470326-134
Electrophoresis Buffer Stock Powders
Buffers Electrophoresis Buffers Loading Buffers
Our premeasured powdered packs offer convenience and will change the way you prepare buffers in your lab.
  • Convenient, premeasured packs make 500 ml or 1 L of 1X solution
  • Convenient dispensing box
  • Add pouch contents to water and stir to dissolve
  • No dilutions of stock solutions needed
Ward's TBE Buffer: Best for gel preparation and as a running buffer for electrophoresis of nucleic acids. TBE works best for resolution of smaller fragments (<2kb). Its higher buffering capacity makes it useful for longer runs. The final concentration of a 1X working solution is 89mM Tris, 89mM boric acid, 2mM EDTA.
Ward's TAE Buffer: Best for recovering DNA from gels and when downstream processing involves enzymatic reactions. Final concentration of a 1X working solution is 40mM Tris, 20mM acetic acid, 1mM EDTA.
Ward's Tris-Glycine-SDS Buffer: One of the most common running buffers used for SDS-PAGE. A 1X solution contains 25mM Tris, 192mM Glycine and 0.1% SDS.